Learn More
CD314 (NKG2D) Rat anti-Mouse, PE-eFluor 610, Clone: CX5, eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 61-5882-80
Code : NEW
Additional Details : Weight : 0.09500kg
Product Code. 15570527
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
The CX5 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1+ thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells.
Specifications
Specifications
CD314 (NKG2D)
CX5
PE-eFluor 610
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
KLRK1
IgG1, kappa
RUO
Primary
Murine