missing translation for 'onlineSavingsMsg'
Learn More

CD314 (NKG2D), PerCP-eFluor™ 710, clone: 1D11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-5878-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15529296

  • £77.01 / 25 tests


EU Stock 2
Add to basket

Description

Description

Description: The 1D11 monoclonal antibody reacts with the human NKG2D, a 42kDa lectin-like molecule expressed by NK cells, gamma delta T cells, CD8+ T cells, and some CD4+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gamma delta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells.Applications Reported: This 1D11 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 1D11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD314 (NKG2D)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P26718
CD314, NK cell receptor D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, killer cell lectin-like receptor subfamily K member 1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
1D11
PerCP-eFluor 710
KLRK1
Conjugated
KLRK1
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
22914
Videos

For Research Use Only.