missing translation for 'onlineSavingsMsg'
Learn More

CD314 (NKG2D), FITC, clone: 1D11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5878-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15546576

  • £159.14 / 100 tests


EU Stock 1
Add to basket

Description

Description

Description: The 1D11 monoclonal antibody reacts with the human NKG2D, a 42kDa lectin-like molecule expressed by NK cells, gamma delta T cells, CD8+ T cells, and some CD4+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gamma delta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells.Applications Reported: The 1D11 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 1D11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD314 (NKG2D)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P26718
CD314, NK cell receptor D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, killer cell lectin-like receptor subfamily K member 1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
1D11
FITC
KLRK1
Conjugated
KLRK1
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
22914
Videos

For Research Use Only.