Learn More
CD314 (NKG2D) Monoclonal Antibody (1D11), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-5878-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The 1D11 monoclonal antibody reacts with the human NKG2D, a 42 kDa lectin-like molecule expressed by NK cells, gamma delta T cells, CD8+ T cells, and some CD4+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gamma delta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells. Applications Reported: The 1D11 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining of frozen tissue sections. It has also been reported in inhibition of ligand binding in in vitro assays. Please use Functional Grade Purified 1D11 (cat. 16-5878) in functional assays. (Fluorochrome-conjugated 1D11 is recommended for use in flow cytometric analysis.). Applications Tested: The 1D11 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. NK cells can regulate specific humoral and cell-mediated immunity, and preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The NK gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.Specifications
CD314 (NKG2D) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P26718 | |
Klrk1 | |
Affinity chromatography | |
RUO | |
22914 | |
4° C | |
Liquid |
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Neutralization | |
1D11 | |
Unconjugated | |
Klrk1 | |
CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.