missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD314 (NKG2D), clone: 1D11, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-5878-80
Code : Z2
Additional Details : Weight : 0.01000kg
Description
The 1D11 monoclonal antibody reacts with the human NKG2D, a 42kDa lectin-like molecule expressed by NK cells, gamma delta T cells, CD8+ T cells, and some CD4+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gamma delta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells.Specifications
CD314 (NKG2D) | |
1D11 | |
Unconjugated | |
Purified | |
Murine | |
25μg | |
Store at 2-8°C. | |
Monoclonal |
Immunoprecipitation | |
0.5mg/mL | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
KLRK1 | |
IgG1 | |
RUO | |
Primary | |
Human |
For Research Use Only.