missing translation for 'onlineSavingsMsg'
Learn More

CD314 (NKG2D), Biotin, clone: A10, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5872-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15279879

  • £124.41 / 100µg


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Description

Description

Description: The A10 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1(+) thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells. A10 and C7 antibodies detect NK cells from all inbred strains of mice tested so far. In redirected lysis assays, A10 is reported to stimulate NK cells to kill Daudi targets. In addition, immobilized A10 is reported to induce production of GM-CSF by NK clones and LAK cells through a CD16 (FC gamma RIII) concomitant engagement. A10 and another hamster anti-mouse NKG2D (clone C7) compete with each other for binding to transfected cells by flow cytometric analysis, suggesting that they may bind to similar epitopes or block each other by steric hindrance. C7 (neutralizing) and A10 (activating) also exhibit different functional properties. Expression of the NKG2D antigen on mouse peripheral NK and NKT cells can be detected by flow cytometric analysis using mAb CX5 with much brighter intensity.Applications Reported: This A10 antibody has been reported for use in flow cytometric analysis.Applications Tested: This A10 antibody has been tested by flow cytometric anlysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD314 (NKG2D)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O54709
CD314, NK cell receptor D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, killer cell lectin-like receptor subfamily K member 1
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
A10
Biotin
Klrk1
Conjugated
Klrk1
IgG
100μg
Store at 2-8°C. Do not freeze.
Primary
27007
Videos

For Research Use Only.