Learn More
CD31 (PECAM-1) Monoclonal Antibody (390), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£82.50 - £166.00
Specifications
Antigen | CD31 (PECAM-1) |
---|---|
Clone | 390 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15331710
|
Affymetrix eBioscience
48-0311-80 |
25 μg |
£82.50
25µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15341710
|
Affymetrix eBioscience
48-0311-82 |
100 μg |
£166.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 390 monoclonal antibody reacts with mouse CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140 kDa surface protein is expressed by endothelial cells and at low levels on all leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction. Applications Reported: This 390 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 390 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD31 (PECAM-1) Monoclonal antibody specifically detects CD31 (PECAM-1) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD31 (PECAM-1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Pecam1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Pecam1 |
390 | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
Q08481 | |
18613 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.