missing translation for 'onlineSavingsMsg'
Learn More

CD31 (PECAM-1), Biotin, clone: 390, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0311-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209529

  • £49.78 / 50µg


EU Stock 3
Add to basket

Description

Description

Description: The 390 monoclonal antibody reacts with mouse CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140kDa surface protein is expressed by endothelial cells and at low levels on all leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction.Applications Reported: The 390 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 390 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD31 (PECAM-1)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q08481
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
390
Biotin
Pecam1
Conjugated
Pecam1
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze.
Primary
18613
Videos

For Research Use Only.