missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine CD31/PECAM-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | CD31/PECAM-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Porcine samples. It is validated for Western Blot.Specifications
CD31/PECAM-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Porcine | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
PECAM1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q95242 | |
5175 | |
Mouse myeloma cell line NS0-derived recombinant porcine CD31/PECAM-1 Gln28-Lys602 Accession # Q95242 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects porcine CD31/PECAM-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human CD31 and recombinant mouse CD31 is observed. |
For Research Use Only