Learn More
CD31/PECAM-1 (Endothelial Cell Marker) Rabbit Polyclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD31/PECAM-1 (Endothelial Cell Marker), Rabbit Polyclonal Antibody.
£165.20 - £565.00
Specifications
Antigen | CD31 (Endothelial Cell Marker) |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12662787
|
Epredia
RB-10333-P1 |
Concentrated | 500 μL |
£347.00
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12642787
|
Epredia
RB-10333-P |
Concentrated | 1 mL |
£565.00
1mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12652787
|
Epredia
RB-10333-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12682787
|
Epredia
RB-10333-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Beschreibung
CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion. Antibody to CD31 is of value in the study of benign and malignant vascular tumors. Staining for CD31 has also been used to measure angiogenesis, which reportedly predicts tumor recurrence.Host Species: Rabbit
Species Reactivity: Human. Others not tested.
Epitope: C-terminal
Immunogen: Synthetic peptide corresponding to C-terminus of mouse CD31 protein
Molecular Weight: 100 and 130 kDa
Positive Control: Tonsil, Placenta
Cellular Localization: Endothelial cells in blood vessel of tonsil
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Spezifikation
CD31 (Endothelial Cell Marker) | |
Polyclonal | |
Rabbit | |
Cancer and Tumor Biology | |
Synthetic peptide corresponding to C-terminus of mouse CD31 protein | |
Tonsil |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Primary |