Learn More
Epredia Monoclonal antibody, anti-CD31/PECAM-1 (Endothelial Cell Marker) Ab-1, host mouse, clone JC/70A (Workshop V), isotype IgG1/k, species reactivity human (Hu), applications IF, IHC (P) Thermo Scientific LabVision
Brand:  Epredia MS-353-R1
Monoclonal antibody, anti-CD31/PECAM-1 (Endothelial Cell Marker) Ab-1, host mouse, clone JC/70A (Workshop V), isotype IgG1/k, species reactivity human (Hu), applications IF, IHC (P) Thermo Scientific LabVision
Code : LD
Additional Details : Weight : 1.00000kg
Product Code. 12691127