missing translation for 'onlineSavingsMsg'
Learn More

CD309 (FLK1), PE, clone: Avas12a1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5821-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15298839

  • £99.63 / 50µg


EU Stock 1
Add to basket

Description

Description

Description: The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult.Applications Reported: The Avas12a1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The Avas12a1 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD309
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P35918
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2, CD309, KDR
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
Avas12a1
PE
Kdr
Conjugated
Kdr
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16542
Videos

For Research Use Only.