missing translation for 'onlineSavingsMsg'
Learn More

CD309 (FLK1), Biotin, clone: Avas12a1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5821-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15279869

  • £180.80 / 500µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult.Applications Reported: The Avas12a1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The Avas12a1 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD309
Avas12a1
Biotin
Conjugated
Rat
500μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Flk-1, VEGF-R2, VEGFR2, Ly-73, Ly73, KDR
IgG2a
RUO
Primary
Murine
Videos