missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD309 (FLK1), Alexa Fluor 700, clone: Avas12a1, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 56-5821-81
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult.Applications Reported: This Avas12a1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This Avas12a1 antibody has been tested by flow cytometric analysis of bEND3 endothelial cell line. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
CD309 (FLK1) | |
Monoclonal | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P35918 | |
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2, CD309, KDR | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Murine |
Flow Cytometry | |
Avas12a1 | |
Alexa Fluor 700 | |
Kdr | |
Conjugated | |
Kdr | |
IgG2a, kappa | |
50μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
16542 |
For Research Use Only.