missing translation for 'onlineSavingsMsg'
Learn More

CD307e (FcRL5), eFluor 660, clone: 509F6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-3078-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15569926

  • £81.61 / 25 tests


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Description: The 509F6 monoclonal antibody reacts with human CD307e which is also known as Fc receptor-like 5, FcRL5, FcRH5, or IRTA2. CD307e is a surface protein expressed selectively on naive mature B cells and on bone marrow plasma cells, and binds to all IgG isotypes. CD307e functions as an inhibitory coreceptor on mature B cells in humans via the recruitment of the phosphatase SHP-1 to its two immunoreceptor tyrosine-based inhibitory motifs (ITIM). CD307e can also exist in soluble form and high levels of soluble CD307e have been detected in the blood of patients with multiple myeloma, chronic lymphocytic leukemia, and mantle cell lymphoma. Because surface-bound CD307e is also highly-expressed on malignant B cells from multiple myeloma, chronic lymphocytic leukemia, and mantle cell lymphoma patients, CD307e may be a useful therapeutic target for the treatment of these cancers.Applications Reported: This 509F6 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 509F6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD307e (FcRL5)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q96RD9
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820, Fc receptor-like protein 5, fc receptor homolog 5, fcR-like protein 5, immune receptor translocation-associated protein 2, immunoglobulin superfamily receptor translocation associated 2 IRTA2
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
509F6
eFluor 660
FCRL5
Conjugated
FCRL5
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
83416
Videos

For Research Use Only.