missing translation for 'onlineSavingsMsg'
Learn More

CD307d (FcRL4), PerCP-eFluor™ 710, clone: 413D12, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-3079-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15549236

  • £90.11 / 25 tests


Estimated Shipment date
from Supplier 01-06-2018
Add to basket

Description

Description

Description: The 413D12 monoclonal antibody reacts with human CD307d, which is also known as FcRL4, Fc receptor-like 4, FcRH4, IRTA4, or IGFP2. CD307d is an inhibitory receptor that is highly expressed on a subset of memory B cells resident in epithelial tissues and marginal zones of mucosal lymphoid tissues such as tonsils. CD307d has been shown to recruit the phosphatase SHP-1, thereby acting as an inhibitory receptor in B cells. CD307d is homologous to FcgammaRI and specifically binds IgA. To date, CD307d is the only known inhibitory receptor for IgA. Expression of CD307d is increased on memory B cells from patients with chronic infections such as HIV, suggesting that enhanced expression of inhibitory receptors, such as CD307d, may contribute to the inefficiency of antibody responses against HIV.Applications Reported: This 413D12 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 413D12 antibody has been pre-titrated and tested by flow cytometric analysis of human tonsil tissue or transfected 293T cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD307d (FcRL4)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q96PJ5
FCRH4, IGFP2, IRTA1, MGC150522, MGC150523
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
413D12
PerCP-eFluor 710
FCRL4
Conjugated
FCRL4
IgG2b, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
83417
Videos

For Research Use Only.