missing translation for 'onlineSavingsMsg'
Learn More

CD305 (LAIR1), PE, clone: 113, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-3051-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15258739

  • £124.41 / 100µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: CD305 or Leukocyte-associated Ig-like receptor-1 (LAIR-1) is a 46 kD cell surface molecule that functions as an inhibitory collagen-receptor on the majority of cells of the immune system. The mouse LAIR-1 molecule is expressed primarily on monocytes/macrophages and also on T cells, NK cells and dendritic cells. Unlike human B cells , mouse B cells do not express LAIR-1. The cytoplasmic tail of mouse LAIR-1 can be phosphorylated leading to the recruitment of SH2-containing tyrosine phosphatase(SHP)-2 and C-terminal Src kinase.Applications Reported: This 113 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 113 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD305 (LAIR-1)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q8BG84
LAIR1, leukocyte-associated immunoglobulin-like receptor 1, CD305, LAIR-1, leukocyte-associated Ig-like receptor 1
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
113
PE
Lair1
Conjugated
Lair1
IgG
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
52855
Videos

For Research Use Only.