missing translation for 'onlineSavingsMsg'
Learn More

CD304 (Neuropilin-1), PerCP-eFluor™ 710, clone: 3DS304M, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-3041-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15509236

  • £89.66 / 25µg


EU Stock 3
Add to basket

Description

Description

Description: The monoclonal antibody 3DS304M recognizes mouse CD304 (Neuropilin-1), a type 1 transmembrane glycoprotein present on the surface of multiple cell types, including: regulatory T cells (Treg), NKT cells, DC, certain types of stem cells, neurons, endothelial cells, and some neoplastic cells. Neuropilin-1, in complex with plexins, serves as a co-receptor for type 3 semaphorins in growing neurons. It is also involved in the process of angiogenesis being a part of a functional receptor for VEGF in endothelial cells. In mice, Neuropilin-1 is expressed on thymus-derived natural Treg but not peripherally induced Treg. In addition, transient expression on recently activated non-regulatory T cells has been observed. Neuropilin-1 has been used as a marker of recent thymic emigrants in the mouse iNKT cell population. It has been shown that Neuropilin-1 forms a complex with TGF beta receptors, activating the latent form of TGF beta (LAP-TGF beta 1) and augmenting canonical Smad2/3 signaling.This 3DS304M antibody will recognize formaldehyde-fixed as well as methanol-treated epitopes.Applications Reported: This 3DS304M antibody has been reported for use in flow cytometric analysis.Applications Tested: This 3DS304M antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD304 (Neuropilin-1)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P97333
BDCA4, CD304, NP1, NRP, VEGF165R, neuropilin-1, transmembrane receptor, vascular endothelial cell growth factor 165 receptor
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
3DS304M
PerCP-eFluor 710
Nrp1
Conjugated
Nrp1
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
18186
Videos

For Research Use Only.