missing translation for 'onlineSavingsMsg'
Learn More

CD304 (Neuropilin-1), eFluor 450, clone: TNKUSOHA, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-3049-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15559736

  • £94.27 / 25 tests


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The TNKUSOHA monoclonal antibody recognizes human CD304, which is also known as BDCA-4 and Neuropilin-1. Like CD303, the CD304 antigen is expressed on human plasmacytoid dendritic cells in normal blood and bone marrow. CD304 can also be detected on T follicular helper cells in inflamed tonsils, as well as on some B-cell acute lymphoblastic leukemias. In addition, CD304 is expressed on numerous non-hematopoietic cell types, including neurons, endothelial, and tumor cells. Plasmacytoid dendritic cells play a key role in the innate immune response against viral pathogens and tumors.Applications Reported: This TNKUSOHA antibody has been reported for use in flow cytometric analysis.Applications Tested: This TNKUSOHA antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD304
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O14786
BDCA4, CD304, NP1, NRP, VEGF165R, neuropilin-1, transmembrane receptor, vascular endothelial cell growth factor 165 receptor
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
TNKUSOHA
eFluor 450
NRP1
Conjugated
NRP1
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
8829
Videos

For Research Use Only.