Learn More
CD304 (Neuropilin-1) Monoclonal Antibody (TNKUSOHA), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£300.00
Specifications
Antigen | CD304 (Neuropilin-1) |
---|---|
Clone | TNKUSOHA |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | eFluor 450 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15569736
|
Affymetrix eBioscience
48-3049-42 |
100 Tests |
£300.00
100 tests |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The TNKUSOHA monoclonal antibody recognizes human CD304, which is also known as BDCA-4 and Neuropilin-1. Like CD303, the CD304 antigen is expressed on human plasmacytoid dendritic cells in normal blood and bone marrow. CD304 can also be detected on T follicular helper cells in inflamed tonsils, as well as on some B-cell acute lymphoblastic leukemias. In addition, CD304 is expressed on numerous non-hematopoietic cell types, including neurons, endothelial, and tumor cells. Plasmacytoid dendritic cells play a key role in the innate immune response against viral pathogens and tumors. Applications Reported: This TNKUSOHA antibody has been reported for use in flow cytometric analysis. Applications Tested: This TNKUSOHA antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD304 (Neuropilin-1) Monoclonal antibody specifically detects CD304 (Neuropilin-1) in Human samples. It is validated for Flow Cytometry.Specifications
CD304 (Neuropilin-1) | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
8829 | |
IgG1 κ | |
Affinity chromatography | |
NRP1 |
TNKUSOHA | |
Monoclonal | |
Liquid | |
RUO | |
O14786 | |
NRP1 | |
Primary | |
Antibody |
For Research Use Only.