missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD304 (Neuropilin-1), eFluor 450, clone: TNKUSOHA, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 48-3049-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The TNKUSOHA monoclonal antibody recognizes human CD304, which is also known as BDCA-4 and Neuropilin-1. Like CD303, the CD304 antigen is expressed on human plasmacytoid dendritic cells in normal blood and bone marrow. CD304 can also be detected on T follicular helper cells in inflamed tonsils, as well as on some B-cell acute lymphoblastic leukemias. In addition, CD304 is expressed on numerous non-hematopoietic cell types, including neurons, endothelial, and tumor cells. Plasmacytoid dendritic cells play a key role in the innate immune response against viral pathogens and tumors.Applications Reported: This TNKUSOHA antibody has been reported for use in flow cytometric analysis.Applications Tested: This TNKUSOHA antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
CD304 | |
Monoclonal | |
5μl/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
O14786 | |
BDCA4, CD304, NP1, NRP, VEGF165R, neuropilin-1, transmembrane receptor, vascular endothelial cell growth factor 165 receptor | |
Murine | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
TNKUSOHA | |
eFluor 450 | |
NRP1 | |
Conjugated | |
NRP1 | |
IgG1, kappa | |
100 Tests | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
8829 |
For Research Use Only.