missing translation for 'onlineSavingsMsg'
Learn More

CD304 (Neuropilin-1) Mouse anti-Human, Alexa Fluor® 700, Clone: TNKUSOHA, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-3049-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15560357

  • £92.25 / 25 tests

Estimated Shipment
04-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Neuropilin 1 is a 923 amino acid containing membrane bound coreceptor to a tyrosine kinase receptor belonging to the neuropilin family with two CUB domains, two F5/8 type C domains and a MAM domain. Neuropilin 1 forms a heterodimer with NRP2 (Neuropilin 2) and is known to bind to plexin (PLXNB1) to form semaphorin receptors. Expressed mostly in the nervous system this receptor is implicated in a wide range of functions including in the development of the cardiovascular system, angiogenesis, repulsive axon guidance, formation of certain neuronal circuits, cell survival, control of neuronal migration and organogenesis outside the nervous system. This semaphorin receptor functions as receptors for both the semaphorins and vascular endothelial growth factor and is also a regulator of VEGF-induced angiogenesis. Neuropilin 1 may be involved in aggressive advanced prostate carcinoma and in the progression of breast cancer.
Specifications

Specifications

CD304 (Neuropilin-1)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O14786
BDCA4, CD304, NP1, NRP, VEGF165R, neuropilin-1, transmembrane receptor, vascular endothelial cell growth factor 165 receptor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
TNKUSOHA
Alexa Fluor™ 700
NRP1
Liquid
NRP1
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
8829
Videos
Documents

For Research Use Only.