missing translation for 'onlineSavingsMsg'
Learn More

CD30, Functional Grade, clone: mCD30.1, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-0301-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15228427

  • £100.59 / 100Āµg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The mCD30.1 monoclonal antibody reacts with mouse CD30, a 105-120kDa member of the TNFR superfamily. CD30 is not detectable on splenocytes, but can be upregulated on T cells stimulated in vitro through TCR with a peak expression around day 4-5 post-activation. CD30 is expressed by both CD8^+ and CD4^+ activated cells. It is suggested that CD30 regulates cytokine secretion by T cells and has a role in T cell development.Applications Reported: The mCD30.1 antibody has been reported for use in flow cytometric analysis. It has also been reported in in vitro functional studies.Applications Tested: The mCD30.1 antibody has been tested by flow cytometric analysis of resting and activated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD30
Monoclonal
1mg/mL
PBS with no preservative; pH 7.2
Q60846
CD30 antigen, CD30L receptor, Ki-1 antigen, cytokine receptor CD30, lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryĆ¾Functional Assay
mCD30.1
Functional Grade
Tnfrsf8
Purified
Tnfrsf8
IgG
100μg
Store at 2-8°C.
Primary
21941
Videos

For Research Use Only.