missing translation for 'onlineSavingsMsg'
Learn More

CD3, APC, clone: eBioG4.18 (G4.18), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0030-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15320550

  • £43.95 / 25µg


Estimated Shipment
30-01-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBioG4.18 monoclonal antibody recognizes the rat CD3 protein. CD3 is a critical component of the T-cell receptor (TCR) and is a marker of the T-cell lineage. CD3 is a complex of several subunits and, upon binding of a TCR ligand, participates in transduction of signals from the TCR to the nucleus which results in a variety of cellular responses including transcription of IL-2 and clonal expansion. \i{in vitro\i}, immobilized G4.18 monoclonal antibody has been demonstrated to activate T cells, while soluble G4.18 inhibited allogeneic mixed-lymphocyte proliferative responses and cell-mediated cytotoxicity to allogeneic target cells. \i{in vivo\i}, G4.18 was demonstrated to induce long-term specific tolerance to an organ allograft.Applications Reported: This G4.18 antibody has been reported for use in flow cytometric analysis.Applications Tested: This G4.18 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD3
eBioG4.18 (G4.18)
APC
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
T3
IgG3
RUO
Primary
Rat
Videos