missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Chimpanzee, Human, PerCP-eFluor™ 710, Clone: SK7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0036-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15589056

  • £87.25 / 25 tests

Estimated Shipment
07-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. The CD3 subunit complex which is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. The genes encoding the CD3 epsilon, gamma and delta polypeptides are located on chromosome 11. Defects in the CD3 gene are associated with CD3 immunodeficiency.
Specifications

Specifications

CD3
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P20963, P04234, P07766, P09693
FLJ18683; T3E; TCRE; HCD3
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Chimpanzee, Human
Flow Cytometry
SK7
PerCP-eFluor™ 710
CD3D
Liquid
CD247, CD3D, CD3E, CD3G
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
451584, 451585, 742330, 748272, 915, 916, 917, 919
Videos
SDS
Documents

For Research Use Only.