missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Chimpanzee, Human, PerCP-eFluor™ 710, Clone: SK7, eBioscience™
Mouse Monoclonal Antibody
£89.75 - £227.00
Specifications
Antigen | CD3 |
---|---|
Clone | SK7 |
Target Species | Chimpanzee, Human |
Conjugate | PerCP-eFluor™ 710 |
Isotype | IgG1, kappa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15589056
|
Affymetrix eBioscience
46-0036-41 |
25 Tests |
£89.75
25 tests |
Estimated Shipment 26-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15599056
|
Affymetrix eBioscience
46-0036-42 |
100 Tests |
£227.00
100 tests |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. The CD3 subunit complex which is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. The genes encoding the CD3 epsilon, gamma and delta polypeptides are located on chromosome 11. Defects in the CD3 gene are associated with CD3 immunodeficiency.Specifications
CD3 | |
Chimpanzee, Human | |
IgG1, kappa | |
Monoclonal | |
Liquid | |
Antibody | |
4° C, store in dark, DO NOT FREEZE! |
SK7 | |
PerCP-eFluor™ 710 | |
Flow Cytometry | |
Primary | |
Monoclonal | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
RUO |
For Research Use Only.