missing translation for 'onlineSavingsMsg'
Learn More

CD3, PE-Cyanine5, clone: UCHT1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 15-0038-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15527706

  • £162.30 / 100 tests


EU Stock 17
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The UCHT1 monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR via immobilized UCHT1 initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.Applications Reported: The UCHT1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This UCHT1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD3
UCHT1
PE-Cyanine5
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Leu-4, T3
IgG1
RUO
Primary
Human
Videos