missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, FITC, Clone: HIT3a, eBioscience™

Mouse Monoclonal Antibody

£50.25 - £120.00

Specifications

Antigen CD3
Clone HIT3a
Target Species Human
Conjugate FITC
Isotype IgG2a, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15556306
View Documents
Affymetrix eBioscience
11-0039-41
25 Tests £50.25
25 tests View alternative products
Save Up to 
»
Estimated Shipment
01-12-2020
Please to purchase this item. Need a web account? Register with us today!
15566306
View Documents
Affymetrix eBioscience
11-0039-42
100 Tests £120.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD3 subunit complex which is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. The genes encoding the CD3 epsilon, gamma and delta polypeptides are located on chromosome 11. Defects in the CD3 gene are associated with CD3 immunodeficiency.
Specifications

Specifications

CD3
Human
IgG2a, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
HIT3a
FITC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.