missing translation for 'onlineSavingsMsg'
Learn More

CD3, eVolve 605, clone: OKT3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 83-0037-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15530737

  • £184.66 / 100 tests


Estimated Shipment date
from Supplier 23-04-2018
Add to basket

Description

Description

Description: The OKT3 monoclonal antibody reacts with an epitope on the epsilon-subunit within the human CD3 complex. The OKT3 antibody has been reported to have potent immunosuppressive properties in vivo and has been proven effective in the treatment of renal, heart and liver allograft rejection. The CD3 subunits, gamma, delta, and epsilon chains, are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.Applications Reported: This OKT3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This OKT3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL per test. A test is defined as the amount of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.Laser/Filter Recommendation: eVolve™ 605 can be optimally excited with laser lines from 325-405 nm, and emits near 610 nm. We recommend using the violet (405 nm) laser line for excitation and a 595LP with a 605/40 or 610/20 bandpass filter, or equivalent, for detection.Fixation Recommendation: Antibodies conjugated to eVolve™ are compatible with eBioscience's fixation and permeabilization buffers.Excitation: 350-590 nm; Emission: 605 nm; Laser: Ultraviolet Laser, Violet LaserStorage and handling: For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.
Specifications

Specifications

CD3
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P20963, P09693, P07766, P04234
FLJ18683, T3E, TCRE
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
OKT3
eVolve 605
CD3E
Conjugated
CD247, CD3D, CD3E, CD3G
IgG2a, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary
915, 916, 917, 919
Videos

For Research Use Only.