missing translation for 'onlineSavingsMsg'
Learn More

CD3, Functional Grade, clone: UCHT1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-0038-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15298347

  • £68.65 / 50µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The UCHT1 monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR via immobilized UCHT1 initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.Applications Reported: The UCHT1 antibody has been reported for use in flow cytometric analysis. UCHT1 has also been reported in activation of T cells.Applications Tested: The UCHT1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD3
UCHT1
aqueous buffer, no sodium azide
Leu-4, T3
IgG1
RUO
Primary
Human
Functional Assay
Unconjugated
Purified
Murine
50μg
Store at 2-8°C.
Monoclonal
Videos