missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, BV786, Clone: UCHT1, BD Horizon™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 565491

Code : NEW

Additional Details : Weight : 0.00100kg

Product Code. 15735978

  • £312.00 / 50 tests


Estimated Shipment date
from Supplier 24-01-2019
Add to basket

Description

Description

CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

CD3
UCHT1
Vol. Per test: 5μL
Purified
Murine
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed.
Monoclonal
Flow Cytometry
Brilliant Violet 786
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor
50 Tests
Cell Biology
Primary
Human
Videos

For Reseach Use Only.