missing translation for 'onlineSavingsMsg'
Learn More

CD294 (CRTH2) Rat anti-Human, eFluor 660, Clone: BM16, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-2949-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15599916

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This BM16 monoclonal antibody reacts with human CD294, which is also known as G protein-coupled receptor-44 (GPR-44) or chemoattractant receptor expressed on Th2 cells (CRTH2). CD294 is expressed on Th2 cells, some CD8+ cells, eosinophils, and basophils. Moreover, CD294 binds prostaglandin D2, which promotes leukocyte chemotaxis during allergic responses. CD294 has been reported to mediate prostaglandin D2-mediated suppression of apoptosis of Th2 cells by activating the PI3K pathway.
Specifications

Specifications

CD294 (CRTH2)
BM16
eFluor 660
Conjugated
IgG2a
RUO
Primary
Human
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos
Documents

For Research Use Only.