missing translation for 'onlineSavingsMsg'
Learn More

CD294 (CRTH2) Rat anti-Human, eFluor 660, Clone: BM16, eBioscience™

Rat Monoclonal Antibody

£60.53 - £171.00

Specifications

Antigen CD294 (CRTH2)
Clone BM16
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15509926 View Documents Affymetrix eBioscience
50-2949-42
100 Tests £171.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Please to purchase this item. Need a web account? Register with us today!
15599916 View Documents Affymetrix eBioscience
50-2949-41
25 Tests This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

This BM16 monoclonal antibody reacts with human CD294, which is also known as G protein-coupled receptor-44 (GPR-44) or chemoattractant receptor expressed on Th2 cells (CRTH2). CD294 is expressed on Th2 cells, some CD8+ cells, eosinophils, and basophils. Moreover, CD294 binds prostaglandin D2, which promotes leukocyte chemotaxis during allergic responses. CD294 has been reported to mediate prostaglandin D2-mediated suppression of apoptosis of Th2 cells by activating the PI3K pathway.
Specifications

Specifications

CD294 (CRTH2)
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
BM16
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test

For Research Use Only.