Learn More
CD294 (CRTH2) Rat anti-Human, APC, Clone: BM16, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 17-2949-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This BM16 monoclonal antibody reacts with human CD294, which is also known as G protein-coupled receptor-44 (GPR-44) or chemoattractant receptor expressed on Th2 cells (CRTH2). CD294 is expressed on Th2 cells, some CD8+ cells, eosinophils, and basophils. Moreover, CD294 binds prostaglandin D2, which promotes leukocyte chemotaxis during allergic responses. CD294 has been reported to mediate prostaglandin D2-mediated suppression of apoptosis of Th2 cells by activating the PI3K pathway. Applications Reported: This BM16 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BM16 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
The prostaglandin D2 receptor 2 (PTGDR2), also known as CRTH2, is expressed on Th2 cells and eosinophils and mediates chemotaxis of these cells to PGD2 and is thus thought to be a key receptor mediating eosinophil and Th2 recruitment during allergic responses. However, PTGDR2-null mice showed enhanced eosinophil recruitment into the lung consistent with observations that the PTGDR2-null mice produced significantly higher amounts of interleukin-5 (IL-5) and IL-3. This suggests that PTGDR2 plays a nonredundant role in restricting eosinophilia and allergic response in vivo.Specifications
CD294 (CRTH2) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9Y5Y4 | |
PTGDR2 | |
Affinity chromatography | |
RUO | |
11251 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
BM16 | |
APC | |
PTGDR2 | |
Crth2, Grp45, MGC130436 | |
Rat | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
For Research Use Only.