missing translation for 'onlineSavingsMsg'
Learn More

CD29 (Integrin beta 1), clone: TS2/16, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0299-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15216897

  • £36.94 / 25µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Description: The TS2/16 monoclonal antibody reacts with human CD29, also known as integrin beta 1, an approximately 130kDa single-pass transmembrane glycoprotein. CD29 complexes with one of nine integrin alpha subunits to form the very late antigen (VLA) subfamily of adhesion molecules. Integrin heterodimers containing CD29 are involved in cell-cell and cell-matrix adhesion. CD29 is expressed broadly on lymphocytes and monocytes, with lower levels of expression on granulocytes. The TS2/16 antibody has been found to possess activating activity for beta 1 integrins.Applications Reported: This TS2/16 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining.Applications Tested: This TS2/16 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD29
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P05556
FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB, ITGB1, ITBG1D
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunoprecipitation
TS2/16
Unconjugated
ITGB1
Purified
ITGB1
IgG1, kappa
25μg
Store at 2-8°C.
Primary
3688
Videos

For Research Use Only.