missing translation for 'onlineSavingsMsg'
Learn More

CD289 (TLR9), FITC, clone: M9.D6, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-9093-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15516616

  • £191.71 / 100µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: M9.D6 is generated against a peptide derived from the extracellular portion of mouse TLR9. Predominantly expressed as an intracellular protein, TLR9 is a ∽115-120kDa molecule which mediates response to unmethylated CpG dinucleotides in bacterial DNA. CpG DNA induces a strong T-helper-1-like inflammatory response and the proliferation of TLR9+ B cells. When stimulated with CpG DNA, TLR9-deficient (TLR9-/-) mice lacked splenocyte proliferation, inflammatory cytokine production from macrophages, and dendritic cell maturation, as compared with normal mice. To date, at least ten members of the Toll family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Members of the TLR family are involved in recognition and response to different microbial components including lipoproteins, peptidoglycans, and nucleic acids and play important roles in innate immunity and inflammation. TLR9 is not detected by flow cytometry using this antibody on RBC-lysed mouse splenocytes stained for intracellular TLR9. This may be due to limitations of antigen detection by flow cytometry. Further studies are needed to determine the relationship between mRNA expression and protein detection.Applications Reported: This M9.D6 antibody has been reported for use in intracellular flow cytometric analysis.Applications Tested: This M9.D6 antibody has been tested by intracellular staining and flow cytometric analysis of fixed and permeabilized mouse TLR9 transfected 293T cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD289 (TLR9)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9EQU3
Toll-like Receptor 9, Toll-like receptor 9 protein, UNQ5798/PRO19605
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
M9.D6
FITC
Tlr9
Conjugated
Tlr9
IgG2a
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
81897
Videos

For Research Use Only.