missing translation for 'onlineSavingsMsg'
Learn More

CD289 (TLR9), clone: eB72-1665, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9099-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238147

  • £61.99 / 25µg


EU Stock 5
Add to basket

Description

Description

Description: eB72-1665 is generated against a portion of human toll-like receptor 9 (aa 273-288), a molecule reported to be expressed predominantly intracellularly. TLR9 is a ∽115-120kDa molecule, which mediates response to unmethylated CpG dinucleotides in bacterial DNA. CpG DNA induces a strong T-helper-1-like inflammatory response and the proliferation of TLR9-positive human B cells. When stimulated with CpG DNA, TLR9-deficient (TLR9-/-) mice lacked splenocyte proliferation, inflammatory cytokine production from macrophages, and dendritic cell maturation, as compared with normal mice. To date, at least twelve members of the Toll family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Members of the TLR family are involved in recognition and response to different microbial components including lipoproteins, peptidoglycans, and nucleic acids and play important roles in innate immunity and inflammation. TLR9 is not detected by flow cytometry using this antibody on lysed whole human blood and/or isolated human PBMC stained for cell surface or intracellular TLR9. This may be due to limitations of antigen detection by flow cytometry. Human pDCs matured in the presence of IL-3 have been reported to stain with eB72-1665 by immunofluorescence microscopy (Nat Immunol. 5:190). Human Epithelial Cell lines were also reported to stain with this mAb (J. Immunol. 173: 1219). Further studies are needed to determine the relationship between mRNA expression and protein detection by flow cytometry.Applications Reported: This eB72-1665 antibody has been reported for use in intracellular flow cytometric analysis, immunoprecipitation, and immunoblotting (WB). (Fluorochrome conjugated eB72-1665 is recommended for use in intracellular flow cytometry.) Preliminary in vitro blocking assays suggested that eB72-1665 may not be effective in blocking of CpG binding. Please contact us for further information.Applications Tested: The eB72-1665 antibody has been tested by intracellular flow cytometric analysis of hTLR9 transfected cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD289 (TLR9)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9NR96
Toll-like Receptor 9, Toll-like receptor 9 protein, UNQ5798/PRO19605
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
eB72-1665
Unconjugated
TLR9
Purified
TLR9
IgG2a, kappa
25μg
Store at 2-8°C.
Primary
54106
Videos

For Research Use Only.