missing translation for 'onlineSavingsMsg'
Learn More

CD284 (TLR4), Alexa Fluor 700, clone: HTA125, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-9917-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15550377

  • £87.94 / 25 tests


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HTA125 monoclonal antibody reacts with human Toll-like receptor 4 (TLR4). So far, at least ten members of the Toll family have been identified in humans. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR4 physically associates with another molecule called MD-2, and together with CD14, this complex is responsible for LPS recognition and signaling. TLR4 is expressed by peripheral blood monocytes. HTA125 has been reported to immunoprecipitate human TLR4 (∽100 kDa) from transfected cells. Most TLR cell surface expression, especially TLR1 and TLR4, occurs at low levels on monocytes and at even lower levels on other cell types including granulocytes and immature dendritic cells (iDC). Furthermore, a relatively high degree of variability in TLR surface expression has been reported among normal donors.Applications Reported: This HTA125 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HTA125 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD284 (TLR4)
HTA125
Alexa Fluor 700
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (1.0μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
toll-like receptor 4
IgG2a
RUO
Primary
Human
Videos