missing translation for 'onlineSavingsMsg'
Learn More

CD284 (TLR4) Mouse anti-Human, Alexa Fluor® 700, Clone: HTA125, eBioscience™

Mouse Monoclonal Antibody

£92.25 - £222.00

Specifications

Antigen CD284 (TLR4)
Clone HTA125
Target Species Human
Conjugate Alexa Fluor™ 700
Isotype IgG2a, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15550377
View Documents
Affymetrix eBioscience
56-9917-41
25 Tests
£92.25
25 tests
View alternative products
Save Up to 
»
Estimated Shipment
07-12-2020
Please to purchase this item. Need a web account? Register with us today!
15560377
View Documents
Affymetrix eBioscience
56-9917-42
100 Tests
£222.00
100 tests
View alternative products
Save Up to 
»
Estimated Shipment
07-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor™ 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. TLR4 is a member of the Toll like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. It has been implicated in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria.
Specifications

Specifications

CD284 (TLR4)
Human
IgG2a, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
HTA125
Alexa Fluor™ 700
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO
SDS

For Research Use Only.