Learn More
CD282 (TLR2) Monoclonal Antibody (6C2), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£124.00 - £311.00
Specifications
Antigen | CD282 (TLR2) |
---|---|
Clone | 6C2 |
Concentration | 1 mg/mL |
Applications | Control, Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15258817
|
Affymetrix eBioscience
16-9021-81 |
50 μg |
£124.00
50µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15268817
|
Affymetrix eBioscience
16-9021-83 |
200 μg |
£311.00
200µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 6C2 monoclonal antibody reacts with mouse Toll-like receptor 2 (TLR2). Mouse TLR2 is expressed by the myeloid lineage, including macrophage and dendritic cells in splenocytes and the RAW264.7 cell line. To date, at least twelve members of the Toll family have been identified in human and mouse. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR2 is expressed on the surface of cells and is responsible for distinguishing different pathogens. Applications Reported: 6C2 has been reported for use in flow cytometric analysis. Unpublished preliminary studies indicate that this antibody may inhibit some aspects of monocyte activation through TLR2, however, it may not result in complete blocking of TLR2 function. For optimal flow cytometry staining with this antibody, it is recommended that a directly conjugated 6C2 (FITC, PE, or Biotin) be used in combination with FcR blocking with anti-mouse CD16/CD32 (clone 93, cat. 14-0161). Applications Tested: The 6C2 antibody has been tested by flow cytometric analysis of mouse splenocytes or Raw264.7 cells. This can be used at less than or equal to 1 μg per test.
TLR2 is a member of the Toll-like receptor (TLR) family which play a fundamental role in pathogen recognition and activation of innate immunity. TLR proteins act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors such as Protein Kinase C (PKC) alpha/beta and NF-kappa-B. TLR2 can form heterodimers with either TLR1 or TLR6, and as a heterodimer, can recognize a variety of bacterial and mycoplasma lipoproteins respectively. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. The various TLRs exhibit different patterns of expression. TLR2 is expressed most abundantly in peripheral blood leukocytes, and mediates host response to Gram-positive bacteria and yeast via stimulation of NF-kappaB. TLR2 aids in the recognition of pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. Ten human homologs of TLRs (TLR1-10) have been described. TLR2 is expressed in human cells such as tonsils, microglia, lymph nodes, and appendices, activated B-cells in germinal centers. CD14+ monocytes express the highest level of TLR2 followed by CD15+ granulocytes, CD19+ B-cells, and CD3+ T-cells. The expression of TLR2 on different cell types are regulated by different immune response modifiers. For example, LPS, GM-CSF, IL-1, and IL-10 up regulates TLR2 whereas IL-4, IFN-gamma, and TNF down regulate TLR2 expression in monocytes.Specifications
CD282 (TLR2) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
CD282; Ly105; TIL4; Tlr2; TLR-2; toll like receptor 2; toll/interleukin-1 receptor-like protein 4; Toll-like receptor; toll-like receptor 2; toll-like receptor 2 variant 1; toll-like receptor 2 variant 2; toll-like-receptor 2, exon 1a_; Toll-like-receptor, exon 1b; Toll-like-receptor, exon 1b_; Toll-like-receptor2, exon 1a | |
Tlr2 | |
Primary | |
4° C | |
Tlr2 |
6C2 | |
Control, Flow Cytometry, Functional Assay | |
Functional Grade | |
Rat | |
Mouse | |
Q9QUN7 | |
24088 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.