missing translation for 'onlineSavingsMsg'
Learn More

CD282 (TLR2), PE, clone: mT2.7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9022-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299339

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The mT2.7 monoclonal antibody reacts with mouse Toll-like receptor 2 (TLR2). Mouse TLR2 is expressed by the myeloid lineage, including macrophage and dendritic cells in splenocytes and the RAW264.7 cell line. To date, at least twelve members of the Toll family have been identified in human and mouse. This family of type I transmembrane protein is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR2 is expressed on the surface of cells and is responsible for distinguishing different pathogens.
Specifications

Specifications

CD282 (TLR2)
mT2.7
PE
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
toll-like receptor 2
IgG2a, kappa
RUO
Primary
Murine
Videos

For Research Use Only.