missing translation for 'onlineSavingsMsg'
Learn More

CD282 (TLR2), Biotin, clone: TL2.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9922-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15300120

  • £75.29 / 25µg


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Description

Description

Description: The TL2.1 monoclonal antibody reacts with human Toll-like receptor 2 (TLR2). To date, at least ten members of the Toll family have been identified in human. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR2 is expressed by peripheral blood monocytes and is responsible for distinguishing different pathogens. TL2.1, a blocking antibody, has been used to study the role of TLR-2 as a pattern recognition receptor in microbial lipoprotein/lipopeptide induced cytokine production from human peripheral blood mononuclear cells. TL2.1 has been reported to immunoprecipitate human TLR2 (∽90 kDa) from PBMC and HMEC.Applications Reported: The TL2.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The TL2.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD282 (TLR2)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O60603
TIL4, Toll-like Receptor 2, toll/interleukin 1 receptor-like 4, toll/interleukin-1 receptor-like protein 4
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
TL2.1
Biotin
TLR2
Conjugated
TLR2
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
7097
Videos

For Research Use Only.