missing translation for 'onlineSavingsMsg'
Learn More

CD28 Mouse anti-Human, FITC, Clone: CD28.2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0289-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566366

  • £51.25 / 25 tests

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary’s Disease.
Specifications

Specifications

CD28
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P10747
MGC138290, Tp44, CD28 antigen, T-cell-specific surface glycoprotein CD28
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
CD28.2
FITC
CD28
Liquid
CD28
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
940
Videos
Documents

For Research Use Only.