missing translation for 'onlineSavingsMsg'
Learn More

CD28 Mouse anti-Human, eFluor 450, Clone: CD28.2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0289-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15549656

  • £67.24 / 25 tests


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

The CD28.2 monoclonal antibody reacts with the human CD28 molecule, a 44kDa homodimer expressed by thymocytes, mature T cells and plasma cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent co-stimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells.
Specifications

Specifications

CD28
CD28.2
eFluor 450
Conjugated
IgG1
RUO
Primary
Human
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal