missing translation for 'onlineSavingsMsg'
Learn More

CD28, Biotin, clone: 37.51, eBioscience™

Syrian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0281-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209519

  • £49.99 / 50µg


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Description

Description

Description: The 37.51 monoclonal antibody reacts with the mouse CD28 molecule, a 45kDa homodimer expressed by thymocytes, mature T cells and NK cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent costimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells.Applications Reported: The 37.51 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 37.51 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Note: It has been observed that increased incubations times (30-45 minutes) with the CD28 antibody can enhance the staining obtained.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD28
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P31041
MGC138290, Tp44, CD28 antigen, T-cell-specific surface glycoprotein CD28
Golden Syrian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
37.51
Biotin
Cd28
Conjugated
Cd28
IgG
50μg
Store at 2-8°C. Do not freeze.
Primary
12487
Videos

For Research Use Only.