missing translation for 'onlineSavingsMsg'
Learn More

CD279 (PD-1), PE, clone: RMP1-30, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9981-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229379

  • £137.64 / 100µg


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The RMP1-30 antibody reacts with mouse PD-1 (programmed death-1), a 55kDa member of the Ig superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease in mice. PD-1 is expressed mainly on activated T and B lymphocytes. Two novel B7 Family members have been identified as PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. RMP1-30 does not block the binding of either B7-H1-Ig or B7-DC-Ig to PD-1 transfectants.Applications Reported: The RMP1-30 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RMP1-30 antibody has been tested by flow cytometric analysis of Con A-stimulated mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD279 (PD-1)
RMP1-30
PE
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Pdcd1, Programmed cell death protein 1
IgG2b
RUO
Primary
Murine
Videos