missing translation for 'onlineSavingsMsg'
Learn More

CD279 (PD-1), PE-eFluor 610, clone: eBioJ105 (J105), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-2799-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15580477

  • £93.50 / 25 tests

Estimated Shipment
05-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The J105 monoclonal antibody reacts with the human PD-1 (programmed death-1), a 55kDa member of the CD28 immunoglobulin superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease. PD-1 is expressed predominantly on activated T and B lymphocytes. Two novel members of the B7 family have been identified as the PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these two PD-1 ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells.Costaining experiments suggest that eBioJ105 recognizes a different epitope than MIH4 (cat. 11-9969).Applications Reported: This eBioJ105 (J105) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioJ105 (J105) antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD279 (PD-1)
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q15116
PD1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Rhesus Monkey
Flow Cytometry
eBioJ105 (J105)
PE-eFluor 610
PDCD1
Liquid
PDCD1
IgG1, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
100135775, 5133
Videos
Documents

For Research Use Only.