missing translation for 'onlineSavingsMsg'
Learn More

CD279 (PD-1), clone: J43, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9985-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15228317

  • £170.59 / 500µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The J43 monoclonal antibody reacts with mouse PD-1 (programmed death-1), a 55kDa member of the Ig superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease in mice. PD-1 is expressed mainly on activated T and B lymphocytes. Two novel B7 Family members have been identified as PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. It is reported that J43 inhibits the binding of mouse PD-L1-Ig and mouse PD-L2-Ig to PD-1/BHK transfected cells. When administrated in vivo, both intact and Fab of J43 are reported to enhance contact hypersensitivity and exacerbate acute GVHD similar to transfer of PD-1-deficient cells. Injection of J43 also exacerbates EAE and NOD diabetes as do specific antibodies to mouse PD-L1 and PD-L2.Applications Reported: The J43 antibody has been reported for use in flow cytometric analysis, and immunoprecipitation. It has also been reported for use in in vitro functional assays. (Please use Functional Grade purified J43, cat. 16-9985, in functional assays.).Applications Tested: The J43 antibody has been tested by flow cytometric analysis of mouse Con-A activated spleen cells and mouse PD-1 transfected cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD279 (PD-1)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q02242
PD1
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Immunoprecipitation
J43
Unconjugated
Pdcd1
Purified
Pdcd1
IgG
500μg
Store at 2-8°C.
Primary
18566
Videos

For Research Use Only.