CD279 (PD-1) Armenian Hamster anti-Mouse, Biotin, Clone: J43, eBioscience™

Armenian Hamster Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 13-9985-85

UNSPSC: 12352200

Code: Z2

Additional Details:
Additional Details: Weight: 0.25000kg



Disclaimers: For Research Use Only.

Product Code. 15330150

Quantity Price
1 £ 256.19 / 500µg
Estimated Shipment date
from Supplier 05-05-2017
Add to basket


Description and Specification

Specification

Host Species Armenian Hamster
Antigen CD279 (PD-1)
Isotype IgG
Conjugate Biotin
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Primary or Secondary Primary
Storage Requirements Store at 2-8°C. Do not freeze.
Concentration 0.5mg/mL
Quantity 500μg
Clone J43
Applications Flow Cytometry
Regulatory Status RUO
Species Reactivity Mouse
Monoclonal or Polyclonal Monoclonal

The J43 monoclonal antibody reacts with mouse PD-1 (programmed death-1), a 55kDa member of the Ig superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease in mice. PD-1 is expressed mainly on activated T and B lymphocytes. Two novel B7 Family members have been identified as PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. It is reported that J43 inhibits the binding of mouse PD-L1-Ig and mouse PD-L2-Ig to PD-1/BHK transfected cells. When administrated in vivo, both intact and Fab of J43 are reported to enhance contact hypersensitivity and exacerbate acute GVHD similar to transfer of PD-1-deficient cells. Injection of J43 also exacerbates EAE and NOD diabetes as do specific antibodies to mouse PD-L1 and PD-L2.