Learn More
CD279 (PD-1) Monoclonal Antibody (J43), APC-eFluor™ 780, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 47-9985-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The J43 monoclonal antibody reacts with mouse PD-1 (programmed death-1), a 55 kDa member of the Ig superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease in mice. PD-1 is expressed mainly on activated T and B lymphocytes. Two novel B7 Family members have been identified as PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. It is reported that J43 inhibits the binding of mouse PD-L1-Ig and mouse PD-L2-Ig to PD-1/BHK transfected cells. When administrated in vivo, both intact and Fab of J43 are reported to enhance contact hypersensitivity and exacerbate acute GVHD similar to transfer of PD-1-deficient cells. Injection of J43 also exacerbates EAE and NOD diabetes as do specific antibodies to mouse PD-L1 and PD-L2. Applications Reported: This J43 antibody has been reported for use in flow cytometric analysis. Applications Tested: This J43 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD279 (PD-1) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q02242 | |
Pdcd1 | |
Affinity chromatography | |
RUO | |
18566 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
J43 | |
APC-eFluor 780 | |
Pdcd1 | |
CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
Armenian Hamster | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
For Research Use Only.